Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496161.2 | XP_003496209.1 | 6 - 7 | 667 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |