Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513016.2 | XP_003513064.1 | 3 - 4 | 918 | 69 | |
GenBank mRNA | JI881971.1 | - | 3 - 4 | 940 | 69 | |
JP039088.1 | - | 3 - 4 | 940 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |