Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644252.1 | XP_007642442.1 | 3 - 4 | 539 | 104 | |
XM_007644253.1 | XP_007642443.1 | 4 - 5 | 348 | 104 | ||
GenBank mRNA | JI876960.1 | - | 3 - 4 | 321 | 104 | |
JP060240.1 | - | 3 - 4 | 321 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |