Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497927.1 | XP_003497975.1 | 4 - 5 | 357 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Ion Channel Probe spans exons |