Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noxa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510560.2 | XP_003510608.1 | 5 - 6 | 839 | 71 | |
XM_007651179.1 | XP_007649369.1 | 4 - 5 | 706 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |