Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510959.2 | XP_003511007.1 | 3 - 4 | 677 | 68 | |
XM_007651530.1 | XP_007649720.1 | 2 - 3 | 463 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |