Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pglyrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511290.1 | XP_003511338.1 | 5 - 6 | 716 | 108 | |
XM_007651825.1 | XP_007650015.1 | 5 - 6 | 716 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |