Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oip5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498066.2 | XP_003498114.1 | 3 - 4 | 582 | 97 | |
GenBank mRNA | JP061146.1 | - | 3 - 4 | 511 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |