Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pttg1ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500562.2 | XP_003500610.1 | 4 - 5 | 518 | 88 | |
GenBank mRNA | JI894381.1 | - | 4 - 5 | 351 | 88 | |
JP053506.1 | - | 4 - 5 | 351 | 88 |