Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fstl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501084.2 | XP_003501132.1 | 9 - 10 | 955 | 76 | |
GenBank mRNA | JI875328.1 | - | 4 - 5 | 478 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |