Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hscb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501710.2 | XP_003501758.1 | 4 - 5 | 599 | 105 | |
GenBank mRNA | JI867316.1 | - | 4 - 5 | 596 | 105 | |
JP054176.1 | - | 4 - 5 | 596 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |