Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apobr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514856.2 | XP_003514904.1 | 2 - 3 | 2591 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |