Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007633880.1 | XP_007632070.1 | 5 - 6 | 1813 | 119 | |
XM_007633884.1 | XP_007632074.1 | 5 - 6 | 1503 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |