Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498404.2 | XP_003498452.1 | 2 - 3 | 674 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |