Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506125.2 | XP_003506173.1 | 2 - 3 | 588 | 62 | |
GenBank mRNA | JI886027.1 | - | 2 - 3 | 537 | 62 | |
JP048978.1 | - | 2 - 3 | 537 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Protease 3' Most Recommended Assay Probe spans exons |