Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007620571.1 | XP_007618761.1 | 2 - 3 | 185 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |