Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640791.1 | XP_007638981.1 | 6 - 7 | 1515 | 111 | |
XM_007640793.1 | XP_007638983.1 | 6 - 7 | 1515 | 111 | ||
XM_007640800.1 | XP_007638990.1 | 6 - 7 | 1512 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |