Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515273.1 | XP_003515321.1 | 2 - 3 | 222 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |