Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650498.1 | XP_007648688.1 | 3 - 4 | 367 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |