Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650838.1 | XP_007649028.1 | 3 - 4 | 816 | 84 | |
GenBank mRNA | JI895145.1 | - | 2 - 3 | 182 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |