Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi44l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496444.2 | XP_003496492.2 | 2 - 3 | 582 | 125 | |
XM_007650080.1 | XP_007648270.1 | 2 - 3 | 582 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |