Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509215.2 | XP_003509263.1 | 5 - 6 | 825 | 88 | |
GenBank mRNA | JP056140.1 | - | 5 - 6 | 734 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |