Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647267.1 | XP_007645457.1 | 6 - 7 | 610 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Ion Channel Probe spans exons |