Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653205.1 | XP_007651395.1 | 5 - 6 | 511 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |