Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504063.1 | XP_003504111.1 | 8 - 9 | 876 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |