Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aatk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653957.1 | XP_007652147.1 | 3 - 4 | 595 | 56 | |
XM_007653960.1 | XP_007652150.1 | 3 - 4 | 347 | 56 | ||
XM_007653965.1 | XP_007652155.1 | 3 - 4 | 317 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Protein Kinase Detect Genomic DNA Recommended Assay Probe spans exons |