Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500610.2 | XP_003500658.1 | 2 - 3 | 1514 | 83 | |
XM_007641370.1 | XP_007639560.1 | 3 - 4 | 1427 | 83 | ||
XM_007641371.1 | XP_007639561.1 | 3 - 4 | 1254 | 83 | ||
XM_007641372.1 | XP_007639562.1 | 4 - 5 | 1168 | 83 |