Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gadd45b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506457.2 | XP_003506505.1 | 1 - 2 | 284 | 68 | |
GenBank mRNA | JI877130.1 | - | 1 - 2 | 143 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Detect Genomic DNA Probe spans exons |