Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magohb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510551.2 | XP_003510599.1 | 1 - 2 | 164 | 108 | |
GenBank mRNA | JP038976.1 | - | 1 - 2 | 161 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |