Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100759463 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510717.1 | XP_003510765.1 | 8 - 9 | 609 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |