Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510724.2 | XP_003510772.1 | 5 - 6 | 1004 | 93 | |
GenBank mRNA | JP041866.1 | - | 4 - 5 | 794 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |