Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650957.1 | XP_007649147.1 | 1 - 2 | 317 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Protein Phosphatase Probe spans exons |