Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650957.1 | XP_007649147.1 | 5 - 6 | 907 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Protein Phosphatase 3' Most Probe spans exons |