Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508056.2 | XP_003508104.1 | 1 - 2 | 1129 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |