Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vstm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513979.1 | XP_003514027.1 | 2 - 3 | 365 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |