Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007624893.1 | XP_007623083.1 | 2 - 3 | 613 | 79 | |
GenBank mRNA | JI883587.1 | - | 1 - 2 | 112 | 79 |