Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007624893.1 | XP_007623083.1 | 3 - 4 | 782 | 82 | |
GenBank mRNA | JI883587.1 | - | 2 - 3 | 281 | 82 |