Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497339.1 | XP_003497387.1 | 1 - 2 | 450 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |