Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498383.2 | XP_003498431.1 | 4 - 5 | 330 | 101 | |
XM_007626935.1 | XP_007625125.1 | 4 - 5 | 330 | 101 | ||
XM_007626959.1 | XP_007625149.1 | 4 - 5 | 330 | 101 | ||
GenBank mRNA | JI868499.1 | - | 4 - 5 | 332 | 101 | |
JP060965.1 | - | 4 - 5 | 332 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |