Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btn1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502195.2 | XP_003502243.1 | 3 - 4 | 925 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons |