Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507652.2 | XP_003507700.1 | 3 - 4 | 1154 | 101 | |
XM_007648224.1 | XP_007646414.1 | 3 - 4 | 1154 | 101 | ||
XM_007648225.1 | XP_007646415.1 | 4 - 5 | 965 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |