Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512921.2 | XP_003512969.1 | 3 - 4 | 529 | 73 | |
GenBank mRNA | JI890017.1 | - | 3 - 4 | 497 | 73 | |
JP064314.1 | - | 3 - 4 | 497 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |