Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spcs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007607650.1 | XP_007605840.1 | 2 - 3 | 242 | 101 | |
GenBank mRNA | JP060875.1 | - | 2 - 3 | 239 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Detect Genomic DNA Probe spans exons |