Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bud13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651654.1 | XP_007649844.1 | 9 - 10 | 2569 | 61 | |
GenBank mRNA | JI868955.1 | - | 6 - 7 | 1253 | 61 | |
JP058061.1 | - | 7 - 8 | 1442 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |