Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bud13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651654.1 | XP_007649844.1 | 5 - 6 | 2062 | 108 | |
GenBank mRNA | JI868955.1 | - | 2 - 3 | 746 | 108 | |
JP058061.1 | - | 3 - 4 | 935 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |