Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497186.2 | XP_003497234.1 | 2 - 3 | 233 | 88 | |
XM_007608450.1 | XP_007606640.1 | 2 - 3 | 233 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase Detect Genomic DNA Probe spans exons |