Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pwwp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502214.2 | XP_003502262.2 | 1 - 2 | 46 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |