Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam83f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507468.1 | XP_003507516.1 | 4 - 5 | 1080 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |