Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pten | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510310.2 | XP_003510358.1 | 7 - 8 | 1084 | 96 | |
GenBank mRNA | JI888696.1 | - | 5 - 6 | 633 | 96 | |
JP051053.1 | - | 5 - 6 | 633 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Protein Phosphatase Probe spans exons |