Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exph5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643379.1 | XP_007641569.1 | 1 - 2 | 455 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |